Skip to Content
Merck
All Photos(2)

Key Documents

390011

Sigma-Aldrich

Potassium hexafluoronickelate(IV)

99%

Synonym(s):

Potassium nickel(IV) fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2NiF6
CAS Number:
Molecular Weight:
250.88
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

99%

form

powder

reaction suitability

reagent type: catalyst
core: nickel

SMILES string

[K+].[K+].F[Ni--](F)(F)(F)(F)F

InChI

1S/6FH.2K.Ni/h6*1H;;;/q;;;;;;2*+1;+4/p-6

InChI key

DNNIPLGVOQXCHW-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Precursor for the powerful oxidizing and fluorinating agents NiF4 and NiF3.[1]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zemva, B. et al.
Journal of the American Chemical Society, 117, 10025-10025 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service