Skip to Content
Merck
All Photos(1)

Documents

389943

Sigma-Aldrich

Heptyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6NCO
CAS Number:
Molecular Weight:
141.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.426 (lit.)

bp

75 °C/15 mmHg (lit.)

density

0.876 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCN=C=O

InChI

1S/C8H15NO/c1-2-3-4-5-6-7-9-8-10/h2-7H2,1H3

InChI key

RFXBSYPBSRSQDU-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lajos Nagy et al.
International journal of molecular sciences, 21(4) (2020-02-23)
In this article, the capability of encoding information using a homologous series of monodisperse monomethoxypolyethylene glycols (mPEG), with a number of ethylene oxide units ranging from nEO = 5 to 8, and monodisperse linear aliphatic isocyanates containing a number of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service