Skip to Content
Merck
All Photos(1)

Key Documents

388645

Sigma-Aldrich

Polybutenes

average Mn ~460 by VPO, isobutylene >90 %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2C(CH3)2-]x[-CH2CH(C2H5)-]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~460 by VPO

composition

isobutylene, >90%

viscosity

210-227 cSt(38 °C)(lit.)

transition temp

Tg −69.6 °C
pour point (ASTM D 97) −35 °C

SMILES string

CCC=C.C\C=C\C

InChI

1S/2C4H8/c2*1-3-4-2/h3-4H,1-2H3;3H,1,4H2,2H3/b4-3+;

InChI key

WTOOLIQYCQJDBG-BJILWQEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrophobic material is derivatized to make detergents and anti-rust additives. Adds tack, flexibility and water repellency to adhesives and coatings. Contributes ′cling′ to LLDPE films. Plasticizer for polymers, lubricants and metal-working fluids.

Features and Benefits

Tackiness increases with molecular weight. Some internal double bonds may be present.

Physical form

Terminal vinyl group.

Other Notes

remainder 1- and 2-butenes

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huaiyu Wang et al.
Acta biomaterialia, 5(1), 279-287 (2008-09-02)
Poly(butylene succinate) (PBSu), a novel biodegradable aliphatic polyester with excellent processability and mechanical properties, is a promising substance for bone and cartilage repair. However, it typically suffers from insufficient biocompatibility and bioactivity after implantation into the human body. In this
X Y Wu et al.
The British journal of ophthalmology, 82(5), 569-576 (1998-08-26)
The goal was to compare the biological response of the corneal stroma with three porous materials: a melt blown microfibre web of polybutylene:polypropylene (80:20); a polyester spun laced fabric (polyethylene terephthalate), and an expanded polytetrafluoroethylene. Fifty per cent of each
Syutaro Yamamoto et al.
Journal of oleo science, 56(9), 463-469 (2007-09-28)
Water-soluble metal working fluids are used for processing of aluminum alloy materials. This short article describes properties of new additives in water-soluble metal working fluids for aluminum alloy materials. Many half esters or diesters were prepared from the reactions of
Takanori Tanino et al.
Applied microbiology and biotechnology, 82(1), 59-66 (2008-11-06)
A Candida antarctica lipase B (CALB)-displaying yeast whole-cell biocatalyst was constructed with the integration of the CALB cell-surface display expression cassette in the yeast genome and cell fusion by mating. Lipase hydrolytic activity of the yeast whole-cell biocatalyst subsequently increased
Ibrahim Bulent Buttanri et al.
Ophthalmic plastic and reconstructive surgery, 29(2), 98-100 (2012-12-14)
To evaluate the effect of suturing the silicone rod to the tarsal plate and the suture material used in suturing on surgical success of frontalis suspension for congenital ptosis. Eighty eyes of 56 consecutive patients who underwent frontalis suspension surgery

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service