Skip to Content
Merck
All Photos(2)

Key Documents

380229

Sigma-Aldrich

Iodomethyl methyl ether

contains copper as stabilizer, 95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€109.00

€109.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€109.00

About This Item

Linear Formula:
ICH2OCH3
CAS Number:
Molecular Weight:
171.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€109.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

95%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.546 (lit.)

bp

39 °C/20 mmHg (lit.)

solubility

organic solvents: soluble(lit.)

density

2.03 g/mL at 25 °C (lit.)

functional group

ether
iodo

storage temp.

2-8°C

SMILES string

COCI

InChI

1S/C2H5IO/c1-4-2-3/h2H2,1H3

InChI key

UUSXSNJAQDJKCG-UHFFFAOYSA-N

General description

Iodomethyl methyl ether (Iodomethoxymethane) is a halo ether. Product contains copper as stabilizer. Rotational, centrifugal distortion constants and the nuclear electric quadrupole coupling constant tensor for iodomethyl methyl ether (CH3OCH2I) and its two deuterated species have been measured by microwave spectral studies.[1] It readily undergoes methoxymethylation reaction with phosphorus, carbon, oxygen and nitrogen containing nucleophiles. It can be synthesized by reacting methylal (dimethoxymethane) with iodotrimethylsilane.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodomethyl Methyl Ether
Jung ME, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2007)
Microwave spectrum and nuclear quadrupole coupling constant tensor of iodomethyl methyl ether.
Fujitake M and Hayashi M.
Journal of Molecular Spectroscopy, 128(2), 444-455 (1988)
The carbon-hydrogen bond strength in dimethyl ether and some properties of iodomethyl methyl ether.
Cruickshank FR and Benson SW.
International Journal of Chemical Kinetics, 1(4), 381-390 (1969)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service