Skip to Content
Merck
All Photos(1)

Key Documents

379441

Sigma-Aldrich

Ethyl 5-amino-1-phenyl-4-pyrazolecarboxylate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N3O2
CAS Number:
Molecular Weight:
231.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

98-100 °C (lit.)

solubility

methanol: soluble 25 mg/mL, clear, colorless

SMILES string

CCOC(=O)c1cnn(c1N)-c2ccccc2

InChI

1S/C12H13N3O2/c1-2-17-12(16)10-8-14-15(11(10)13)9-6-4-3-5-7-9/h3-8H,2,13H2,1H3

InChI key

AYJIUOZKKTUKKD-UHFFFAOYSA-N

General description

Ethyl 5-amino-1-phenyl-4-pyrazolecarboxylate is a heterocyclic building block.

Application

Ethyl 5-amino-1-phenyl-4-pyrazolecarboxylate may be used as internal standard for the GC-MS determination of etomidate [ethyl-1-(1-phenylethyl)-1H-imidazole-5-carboxylate] in mouse brain tissue.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Deng et al.
Journal of pharmacological and toxicological methods, 43(1), 73-77 (2000-11-25)
A simple, rapid and reliable method was developed to determine the concentration of etomidate [ethyl-1-(1-phenylethyl)-1 H-imidazole-5-carboxylate] in mouse brain tissue by gas chromatography-mass spectrometry (GC-MS). Ethyl 5-amino-1-phenyl-4-pyrazolecarboxylate was used as internal standard (IS) and liquid-liquid extraction using ethyl ether as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service