Skip to Content
Merck
All Photos(1)

Key Documents

378275

Sigma-Aldrich

2,5-Dimethylresorcinol

95%

Synonym(s):

β-Orcinol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€99.40

€99.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€99.40

About This Item

Linear Formula:
(CH3)2C6H2-1,3-(OH)2
CAS Number:
Molecular Weight:
138.16
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€99.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

95%

bp

277 °C (lit.)

mp

157-162 °C (lit.)

SMILES string

Cc1cc(O)c(C)c(O)c1

InChI

1S/C8H10O2/c1-5-3-7(9)6(2)8(10)4-5/h3-4,9-10H,1-2H3

InChI key

GHVHDYYKJYXFGU-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service