Skip to Content
Merck
All Photos(2)

Key Documents

377120

Sigma-Aldrich

Fullerite

greener alternative

(C60/C70 mixture)

Synonym(s):

Fullerenes, C60/C70 mixture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

InChI

1S/C60/c1-2-5-6-3(1)8-12-10-4(1)9-11-7(2)17-21-13(5)23-24-14(6)22-18(8)28-20(12)30-26-16(10)15(9)25-29-19(11)27(17)37-41-31(21)33(23)43-44-34(24)32(22)42-38(28)48-40(30)46-36(26)35(25)45-39(29)47(37)55-49(41)51(43)57-52(44)50(42)56(48)59-54(46)53(45)58(55)60(57)59

InChI key

XMWRBQBLMFGWIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fullerite is a mixture of C60 and C70 fullerenes. It is a crystalline material with high light absorption and an energy band gap of 1.6 eV. It can be used as an acceptor molecule for the fabrication of bulk heterojunction polymeric solar cells.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mixed C60/C70 based fullerene acceptors in polymer bulk-heterojunction solar cells.
Andersson LM, et al.
Organic Electronics, 13(12), 2856-2864 (2012)
High pressure study of the absorption edge of crystalline C60/C70 mixture.
Dolganov VK, et al.
Solid State Communications, 83(1), 63-65 (1992)
Polymerized fullerite structures.
Nunez-Regueiro M, et al.
Physical Review Letters, 74(2), 278-278 (1995)

Articles

Find various photovoltaic and bioscience-based applications of fullerenes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service