Skip to Content
Merck
All Photos(3)

Key Documents

376876

Sigma-Aldrich

N-Methoxy-N-methyl(triphenylphosphoranylidene)acetamide

98%

Synonym(s):

(N-Methoxymethylaminocarbonylmethylene)triphenylphosphorane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€116.00
25 G
€528.00

€116.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€116.00
25 G
€528.00

About This Item

Linear Formula:
(C6H5)3P=CHCON(OCH3)CH3
CAS Number:
Molecular Weight:
363.39
Beilstein:
4198472
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

€116.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

173-175 °C (lit.)

functional group

amine
phosphine

SMILES string

CON(C)C(=O)C=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C22H22NO2P/c1-23(25-2)22(24)18-26(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21/h3-18H,1-2H3

InChI key

DGBLSOOPDBHHRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Synthesis of electron-deficient alkenes via stereoselective olefination of N-sulfonyl imines
  • Phosphine-catalyzed asymmetric addition reactions
  • Cysteine-catalyzed enantioselective intramolecular Rauhut-Currier reaction
  • Wittig olefination
  • Synthesis of aigialomycin D, a Protein Kinase inhibitor and a potential anticancer agent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service