Skip to Content
Merck
All Photos(1)

Key Documents

374423

Sigma-Aldrich

3-Trimethylsiloxy-1-propyne

98%

Synonym(s):

(Propargyloxy)trimethylsilane, O-(Trimethylsilyl)propargyl alcohol, Trimethyl(2-propynyloxy)silane, Trimethyl(propargyloxy)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH2OSi(CH3)3
CAS Number:
Molecular Weight:
128.24
Beilstein:
2036793
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.408 (lit.)

bp

52 °C/85 mmHg (lit.)

density

0.835 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OCC#C

InChI

1S/C6H12OSi/c1-5-6-7-8(2,3)4/h1H,6H2,2-4H3

InChI key

ZZRPJWCNCLSOLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Trimethylsiloxy-1-propyne is an organosilicon compound. It is reported as silylated acetylenic monomer and its polymerizability has been studied in the presence of combined catalyst iron(III) propanoate-triethylaluminium. N2 laser light-induced aerosol particle formation from 3-trimethylsiloxy-1-propyne (trimethyl(2-propynyloxy)silane) and acrolein has been reported. The infrared laser-photosensitized (SF6) decomposition of trimethyl(2-propynyloxy)silane is reported to afford a multitude of unsaturated hydrocarbons and a solid polydimethylsiloxane phase.

Application

3-Trimethylsiloxy-1-propyne (Trimethyl(2-propynyloxy)silane) may be used in the synthesis of polytrialkylsiloxy-substituted macromolecules. It may be utilized in the preparation of functionalized 4-enynes and 1,4-dienes.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UV laser-induced gas-phase polymerization of trimethyl (propynyloxy) silane.
Pola J and Morita H.
Tetrahedron, 38(44), 7809-7812 (1997)
Journal of Organometallic Chemistry, 309, 1-1 (1986)
IR laser photosensitized decomposition of trimethyl (2-propynyloxy) silane for chemical vapour deposition of polydimethylsiloxane phases.
Urbanova M, et al.
Journal of Analytical and Applied Pyrolysis, 44(2), 219-226 (1992)
Journal of Organometallic Chemistry, 420, 163-163 (1991)
Laser-induced aerosol particle formation from a gaseous mixture of trimethyl (2-propynyloxy) silane and acrolein.
Morita H, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 116(2), 91-95 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service