Skip to Content
Merck
All Photos(1)

Key Documents

374288

Sigma-Aldrich

4-Chlorobenzoyl chloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4COCl
CAS Number:
Molecular Weight:
175.01
Beilstein:
471606
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.578 (lit.)

bp

102-104 °C/11 mmHg (lit.)

mp

11-14 °C (lit.)

density

1.365 g/mL at 20 °C (lit.)

functional group

acyl chloride
chloro

SMILES string

ClC(=O)c1ccc(Cl)cc1

InChI

1S/C7H4Cl2O/c8-6-3-1-5(2-4-6)7(9)10/h1-4H

InChI key

RKIDDEGICSMIJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chlorobenzoyl chloride is an acyl chloride. It reacts with 2-amino-2-seleno-5,5-dimethyl-1,3,2-dioxaphosphorinane to yield the respective N-acyl selenophosphoramides.[1]

Application

4-Chlorobenzoyl chloride may be used in the following studies:
  • Acylation of benzene using different solid acid catalysts such as dodecatungstophosphoric acid (DTPA), DTPA/K-10 clay, K-10, Amberlite, Amberlyst-15, Indion-130, Filtrol-24 clay and sulfated zirconia.[2]
  • Preparation of 1-(4-chlorobenzoyl)-2, 7-dimethoxynaphthalene.[3]
  • Preparation of 4-chlorobenzoyl CoA, via reaction with CoA in KHCO3 buffer.[4]
  • Preparation of 1-aryloxyacetyl-4-(4-chlorobenzoyl)-semicarbazides.[5]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Friedel-Crafts acylation using sulfated zirconia catalyst.
Yadav GD and Pujari AA.
Green Chemistry, 1(2), 69-74 (1999)
A Facile Method to 1, 4-Diacyl Semicarbazides: Syntheses of 1-Aryloxyacetyl-4-(4-Chlorobenzoyl)-Thiosemicarbazides and Semicarbazides.
Wang X, et al.
Synthetic Communications, 30(18), 3405-3411 (2000)
Ryosuke Mitsui et al.
Acta crystallographica. Section E, Structure reports online, 64(Pt 7), o1278-o1278 (2008-01-01)
In the title compound, C(19)H(15)ClO(3), the dihedral angle between the naphthalene ring system and the benzene ring is 72.06 (7)°. The 4-chloro-phenyl group and the carbonyl group are almost coplanar. An inter-molecular C-H⋯O hydrogen bond is formed between an H atom
Grzegorz Cholewinski et al.
Organic & biomolecular chemistry, 7(19), 4095-4100 (2009-09-19)
2-Amino-2-seleno-5,5-dimethyl-1,3,2-dioxaphosphorinane reacts with acyl chlorides (4-chlorobenzoyl chloride or pivaloyl chloride) yielding the respective N-acyl selenophosphoramides. These derivatives do not isomerise to the related selenocarbonyl imides. X-ray study of N-(4-chlorobenzoyl)-2-amino-2-seleno-5,5-dimethyl-1,3,2-dioxaphosphorinane indicates that the selenium atom is placed in the equatorial position.
S D Copley et al.
Applied and environmental microbiology, 58(4), 1385-1387 (1992-04-01)
4-Chlorobenzoate degradation in cell extracts of Acinetobacter sp. strain 4-CB1 occurs by initial synthesis of 4-chlorobenzoyl coenzyme A (4-chlorobenzoyl CoA) from 4-chlorobenzoate, CoA, and ATP. 4-Chlorobenzoyl CoA is dehalogenated to 4-hydroxybenzoyl CoA. Following the dehalogenation reaction, 4-hydroxybenzoyl CoA is hydrolyzed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service