Skip to Content
Merck
All Photos(1)

Key Documents

369071

Sigma-Aldrich

2-Bromo-4,5-difluorophenol

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€40.60

€40.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€40.60

About This Item

Linear Formula:
BrC6H2(F)2OH
CAS Number:
Molecular Weight:
208.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€40.60


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.531 (lit.)

density

1.829 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

Oc1cc(F)c(F)cc1Br

InChI

1S/C6H3BrF2O/c7-3-1-4(8)5(9)2-6(3)10/h1-2,10H

InChI key

FCYZOOHWUOEAOX-UHFFFAOYSA-N

General description

2-Bromo-4,5-difluorophenol is reported to undergo palladium-catalyzed cyclotrimerization to yield hexafluoro- and trimethoxytriphenylene derivatives.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Palladium-Catalyzed Cyclotrimeriza-tion of Arynes: Synthesis of Triphenylenes
PenAa D, et al.
Angewandte Chemie (International Edition in English), 37 (1998)
Yi-Lin Liu et al.
The Journal of organic chemistry, 74(8), 3199-3202 (2009-03-20)
Palladium-catalyzed cocyclotrimerization of allenes with arynes has been developed for selectively synthesizing phenanthrenes. In the presence of [(allyl)PdCl](2) and P(o-tol)(3), a variety of allenes, including internal and terminal allenes, underwent the cocyclotrimerization with arynes to afford the corresponding phenanthrenes in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service