Skip to Content
Merck
All Photos(1)

Key Documents

368539

Sigma-Aldrich

Methylthiomethyl phenyl sulfone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH2SCH3
CAS Number:
Molecular Weight:
202.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

85-87 °C (lit.)

SMILES string

CSCS(=O)(=O)c1ccccc1

InChI

1S/C8H10O2S2/c1-11-7-12(9,10)8-5-3-2-4-6-8/h2-6H,7H2,1H3

InChI key

XBGGZYQBIQAYII-UHFFFAOYSA-N

General description

Methylthiomethyl phenyl sulfone is a sulfone derivative.

Application

Methylthiomethyl phenyl sulfone may be employed:
  • as nucleophilic and electrophilic building block, in the asymmetric synthesis of 6-N-benzoyl-5′-O-benzyl-2′-deoxyadenosine and its anomer[1]
  • in the synthesis of various multidentate methylthio arylethynes (MTA) derivatives[2]
  • as starting reagent in the preparation of methylthio arylbutadiynes (Ar-C=C-C=C-SCH3)[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rigid, conjugated and shaped arylethynes as mediators for the assembly of gold nanoparticles.
Yan H, et al.
Journal of Materials Chemistry, 21(6), 1890-1901 (2011)
Efficient One-Pot Preparation of Methylthio Arylbutadiynes by Double Elimination Protocol.
Su Q, et al.
Synthetic Communications, 43(19), 2648-2655 (2019)
B M Trost et al.
Carbohydrate research, 202, 1-12 (1990-07-15)
An asymmetric synthesis of 6-N-benzoyl-5'-O-benzyl-2'-deoxyadenosine and its a anomer from non-carbohydrate building blocks is achieved in 7 steps. The sequence builds the basic structures using bis(methylthio)methane and methylthiomethyl phenyl sulfone as both nucleophilic and electrophilic building blocks, a feature that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service