Skip to Content
Merck
All Photos(2)

Key Documents

367737

Sigma-Aldrich

Trimethyl(trifluoromethyl)silane solution

0.5 M in THF

Synonym(s):

(Trifluoromethyl)trimethylsilane, Ruppert′s reagent, Ruppert-Prakash reagent, TFMTMS, Trifluoromethyltrimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCF3
CAS Number:
Molecular Weight:
142.19
Beilstein:
4241868
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

vapor pressure

10.98 psi ( 55 °C)
2.8 psi ( 20 °C)

form

liquid

concentration

0.5 M in THF

bp

40 °C

density

0.895 g/mL at 25 °C

SMILES string

C[Si](C)(C)C(F)(F)F

InChI

1S/C4H9F3Si/c1-8(2,3)4(5,6)7/h1-3H3

InChI key

MWKJTNBSKNUMFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nucleophilic addition of the trifluoromethyl group to aldehydes and ketones.
Reactant for:
  • Silver-mediated C-H trifluoromethylation of arenes
  • Preparation of trifluoromethyl ketone analog of L-arginine having contrasting inhibitory activity against human arginase I and histone deacetylase 8
  • Organocatalyzed regio- and enantioselective allylic trifluoromethylation of Morita-Baylis-Hillman adducts
  • Palladium-catalyzed oxidative trifluoromethylation of indoles
  • Preparation of 5-HT1A antagonists

  • Used as difluorocarbene source

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 111, 393-393 (1989)
The Journal of Organic Chemistry, 57, 1124-1124 (1992)
J W Skiles et al.
Journal of medicinal chemistry, 35(4), 641-662 (1992-02-21)
A series of tripeptides possessing trifluoromethyl or aryl ketone residues at P1 were prepared and evaluated both in vitro and in vivo as potential inhibitors of human leukocyte elastase (HLE). Tripeptides containing non naturally occurring N-substituted glycine residues at the
G K Surya Prakash et al.
The Journal of organic chemistry, 71(18), 6806-6813 (2006-08-26)
Organofluorine compounds are becoming increasingly important in different fields, such as material science, agro chemistry, and the pharmaceutical industry. Nucleophilic trifluoromethylation is one of the widely used methods to incorporate a trifluoromethyl moiety into organic molecules. We have carried out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service