Skip to Content
Merck
All Photos(1)

Key Documents

367451

Supelco

N,O-Bis(trimethylsilyl)acetamide

5% in TMCS

Synonym(s):

BSA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
203.43
Beilstein:
1306669
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
Pricing and availability is not currently available.

concentration

5% in TMCS

refractive index

n20/D 1.417 (lit.)

bp

71-73 °C/35 mmHg (lit.)

density

0.832 g/mL at 20 °C (lit.)

SMILES string

C\C(O[Si](C)(C)C)=N/[Si](C)(C)C

InChI

1S/C8H21NOSi2/c1-8(9-11(2,3)4)10-12(5,6)7/h1-7H3/b9-8+

InChI key

SIOVKLKJSOKLIF-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Moiseeva et al.
Cell cycle (Georgetown, Tex.), 14(15), 2408-2421 (2015-06-02)
Expression of oncogenes or short telomeres can trigger an anticancer response known as cellular senescence activating the p53 and RB tumor suppressor pathways. This mechanism is switched off in most tumor cells by mutations in p53 and RB signaling pathways.
Anna Hjelm et al.
Applied and environmental microbiology, 81(2), 726-735 (2014-11-16)
Bacterial ghosts are empty cell envelopes of Gram-negative bacteria that can be used as vehicles for antigen delivery. Ghosts are generated by releasing the bacterial cytoplasmic contents through a channel in the cell envelope that is created by the controlled
Mohammad Saraji et al.
Journal of chromatography. A, 1098(1-2), 30-36 (2005-11-30)
Trace analysis of phenolic compounds in water was performed by coupling single-drop microextraction (SDME) with in-syringe derivatization of the analytes and GC-MS analysis. The analytes were extracted from a 3ml sample solution using 2.5microl of hexyl acetate. After extraction, derivatization
Kazuya Omi et al.
Clinical chemistry, 61(4), 627-635 (2015-02-20)
Small molecules classified as haptens are generally measured by competitive immunoassay, which is theoretically inferior to noncompetitive sandwich immunoassay in terms of sensitivity and specificity. We created a method for developing sandwich immunoassays to measure haptens on the basis of
Anne Burtey et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(11), 4695-4712 (2015-07-30)
Intercellular communication between cancer cells, especially between cancer and stromal cells, plays an important role in disease progression. We examined the intercellular transfer of organelles and proteins in vitro and in vivo and the role of tunneling nanotubes (TNTs) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service