366439
1,1′-Oxalyldiimidazole
technical grade
Synonym(s):
Oxalic acid diimidazolide
About This Item
Recommended Products
grade
technical grade
form
powder
reaction suitability
reaction type: Carbonylations
mp
114-116 °C (lit.)
application(s)
peptide synthesis
SMILES string
O=C(C(=O)n1ccnc1)n2ccnc2
InChI
1S/C8H6N4O2/c13-7(11-3-1-9-5-11)8(14)12-4-2-10-6-12/h1-6H
InChI key
ONRNRVLJHFFBJG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
N-Acylimidazoles and carbonylimidazoles are reactive intermediates used for amino compound acylation, surpassing DCC as coupling reagents.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service