Skip to Content
Merck
All Photos(1)

Key Documents

365157

Sigma-Aldrich

1,1,3,3-Tetramethyl-1,3-diphenyldisilazane

96%

Synonym(s):

1,3-Diphenyl-1,1,3,3-tetramethyldisilazane, DPTMDS

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€210.00

€210.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€210.00

About This Item

Linear Formula:
[C6H5Si(CH3)2]2NH
CAS Number:
Molecular Weight:
285.53
Beilstein:
2738118
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€210.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

96%

form

liquid

refractive index

n20/D 1.538 (lit.)

bp

96-100 °C/0.1 mmHg (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(N[Si](C)(C)c1ccccc1)c2ccccc2

InChI

1S/C16H23NSi2/c1-18(2,15-11-7-5-8-12-15)17-19(3,4)16-13-9-6-10-14-16/h5-14,17H,1-4H3

InChI key

HIMXYMYMHUAZLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,1,3,3-Tetramethyl-1,3-diphenyldisilazane can be used:
  • As a starting material for the synthesis of its substitution products with the tetrachlorides of silicon and tin.[1]
  • To prepare Cu-, Zn- and H2-phthalocyanines using phthalimide.[2]
  • As a pre-column deactivation agent prior to the determination of the fatty acids by GC-MS.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silazanes plus MCl4-substitution vs. rearrangement reactions
Lehnert C, et al.
Chemistry of Heterocyclic Compounds, 42(12), 1574-1584 (2006)
Convenient synthesis of phthalocyanines with disilazanes under mild conditions.
Uchida H, et al.
ARKIVOC (Gainesville, FL, United States), 11, 17-23 (2005)
Dorothee Eibler et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1046, 138-146 (2017-02-10)
Between 2008 and 2011, four polar bears (Ursus maritimus) from the Greenland population swam and/or drifted on ice to Iceland where they arrived in very poor body condition. Body fat resources in these animals were only between 0% and 10%
Simon Hammann et al.
Analytical and bioanalytical chemistry, 407(30), 9019-9028 (2015-10-07)
Triacylglycerols represent the major part (>90%) in most plant oils and have to be eliminated, when the minor compounds such as phytosterols or tocopherols should be analyzed. Here, we used an all liquid-liquid chromatographic technique, countercurrent chromatography (CCC), to fractionate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service