Skip to Content
Merck
All Photos(1)

Key Documents

363871

Sigma-Aldrich

4-Chlorophenyl chloroformate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2C6H4Cl
CAS Number:
Molecular Weight:
191.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

100-102 °C/12 mmHg (lit.)

density

1.365 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC(=O)Oc1ccc(Cl)cc1

InChI

1S/C7H4Cl2O2/c8-5-1-3-6(4-2-5)11-7(9)10/h1-4H

InChI key

RYWGPCLTVXMMHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chlorophenyl chloroformate was used in the preparation of N-hydroxy-N-methylcarbamate 4-chlorophenyl ester.[1]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D S Hamilton et al.
The Journal of biological chemistry, 267(35), 24933-24936 (1992-12-15)
In principle, competitive inhibitors of glyoxalase I that also serve as substrates for the thioester hydrolase glyoxalase II might function as tumor-selective anti-cancer agents, given the role of these enzymes in removing cytotoxic methylglyoxal from cells and the observation that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service