Skip to Content
Merck
All Photos(1)

Documents

363537

Sigma-Aldrich

(S)-(+)-Glycidyl benzyl ether

99%

Synonym(s):

(S)-(+)-2-(Benzyloxymethyl)oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
Beilstein:
5246494
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

optical activity

[α]20/D +5.1°, c = 5 in toluene

density

1.072 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C(OCc1ccccc1)[C@@H]2CO2

InChI

1S/C10H12O2/c1-2-4-9(5-3-1)6-11-7-10-8-12-10/h1-5,10H,6-8H2/t10-/m1/s1

InChI key

QNYBOILAKBSWFG-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(+)-Glycidyl benzyl ether can be used to prepare 1-octadecyl-3-O-benzyl-sn-glycerol, which is a key intermediate for the synthesis of triglycerides containing α-eleostearic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meddy El Alaoui et al.
Chemistry and physics of lipids, 206, 43-52 (2017-06-21)
Lipases are essentially described as sn-1 and sn-3 regio-selective. Actually few methods are available to measure this lipase regio-selectivity, moreover they require chiral chromatography analysis or specific derivations which are discontinuous and time consuming. In this study we describe a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service