Skip to Content
Merck
All Photos(1)

Key Documents

360619

Sigma-Aldrich

3-Bromo-4-methylaniline

98%

Synonym(s):

3-Bromo-p-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH3)C6H3NH2
CAS Number:
Molecular Weight:
186.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

254-257 °C (lit.)

mp

27-30 °C (lit.)

functional group

bromo

SMILES string

Cc1ccc(N)cc1Br

InChI

1S/C7H8BrN/c1-5-2-3-6(9)4-7(5)8/h2-4H,9H2,1H3

InChI key

GRXMMIBZRMKADT-UHFFFAOYSA-N

Application

3-Bromo-4-methylaniline may be used in the synthesis of 1,7-dihalo Tröger′s base isomers.[1] It is suitable for use to investigate the reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations with O2 by ion-trap mass spectrometry.[2]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew B Prendergast et al.
Physical chemistry chemical physics : PCCP, 15(47), 20577-20584 (2013-11-05)
The reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations (m/z 149) with O2 are studied in the gas phase using ion-trap mass spectrometry. Photodissociation (PD) of halogenated precursors gives rise to the target distonic charge-tagged methylphenyl radical whereas collision-induced dissociation
Matthew B Prendergast et al.
Physical chemistry chemical physics : PCCP, 18(6), 4320-4332 (2015-10-29)
Gas-phase product detection studies of o-hydroxyphenyl radical and O2 are reported at 373, 500, and 600 K, at 4 Torr (533.3 Pa), using VUV time-resolved synchrotron photoionisation mass spectrometry. The dominant products are assigned as o-benzoquinone (C6H4O2, m/z 108) and
Halogenation of Troger's base analogues.
Faroughi M, et al.
ARKIVOC (Gainesville, FL, United States), 2, 269-280 (2009)
Annika Schulze et al.
Thrombosis research, 134(1), 174-181 (2014-05-13)
The key feature of heparin-induced thrombocytopenia (HIT) is the production of antibodies (Ab) against the platelet factor 4 (PF4)/heparin complex. These Ab are directed against neoepitopes of the PF4 tetramer, which are induced by the complex formation with heparin. To
Laura Kerosuo et al.
Molecular biology of the cell, 25(3), 347-355 (2013-12-07)
Myc interacting zinc finger protein-1 (Miz1) is a transcription factor known to regulate cell cycle- and cell adhesion-related genes in cancer. Here we show that Miz1 also plays a critical role in neural crest development. In the chick, Miz1 is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service