Skip to Content
Merck
All Photos(1)

Key Documents

360244

Sigma-Aldrich

7-Methyluric acid

98%

Synonym(s):

7-Methyl-2,6,8-trihydroxypurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O3
CAS Number:
Molecular Weight:
182.14
Beilstein:
181629
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

98%

mp

>300 °C (lit.)

SMILES string

CN1C(=O)NC2=C1C(=O)NC(=O)N2

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service