Skip to Content
Merck
All Photos(2)

Key Documents

359491

Sigma-Aldrich

Trimethyl(methylthio)silane

97%

Synonym(s):

(Methylthio)trimethylsilane, Methyl trimethylsilyl sulfide, Trimethylsilyl methyl sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€438.00

€438.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€438.00

About This Item

Linear Formula:
(CH3)3SiSCH3
CAS Number:
Molecular Weight:
120.29
Beilstein:
2035839
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€438.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

110-114 °C (lit.)

density

0.848 g/mL at 25 °C (lit.)

SMILES string

CS[Si](C)(C)C

InChI

1S/C4H12SSi/c1-5-6(2,3)4/h1-4H3

InChI key

JDYCDPFQWXHUDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service