Skip to Content
Merck
All Photos(3)

Key Documents

359408

Sigma-Aldrich

Methoxycarbonylsulfenyl chloride

97%

Synonym(s):

S-Chloro O-methyl thiocarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOSCl
CAS Number:
Molecular Weight:
126.56
Beilstein:
1849908
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.481 (lit.)

bp

133-134 °C (lit.)

density

1.399 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)SCl

InChI

1S/C2H3ClO2S/c1-5-2(4)6-3/h1H3

InChI key

TXJXPZVVSLAQOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methoxycarbonylsulfenyl chloride is a sulfenyl carbonyl compound.[1] Photoinduced fragmentations of methoxycarbonylsulfenyl chloride is investigated in the gaseous phase by using synchrotron radiation and multicoincidence techniques.[2] The molecular structure and conformational properties of methoxycarbonylsulfenyl chloride is studied in the gas and solid phases by gas electron diffraction, low-temperature X-ray diffraction and vibrational spectroscopy.[3] The anomeric and mesomeric effects in methoxycarbonylsulfenyl chloride is reported.[1]

Application

Methoxycarbonylsulfenyl chloride may be used to prepare 3-[(methoxycarbonyl)dithio]-L-alanine, via reaction with L-cysteine.[4] It is suitable for use in the synthesis of 3-2 pyridinyldithio propanoic acid hydrazide (PDPH), an important heterobifunctional crosslinker widely used in bioconjugate techniques for making macromolecule-drug conjugates.[5]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Starkenmann et al.
Journal of agricultural and food chemistry, 59(17), 9457-9465 (2011-08-23)
The L-cysteine derivatives (R)-2-amino-3-(methyldisulfanyl)propanoic acid (S-methylthio-L-cysteine), (R)-2-amino-3-(propyldisulfanyl)propanoic acid (S-propylthio-L-cysteine), (R)-2-amino-3-(1-propenyldisulfanyl)propanoic acid (S-(1-propenylthio)-L-cysteine), and (R)-2-amino-3-(2-propenyldisulfanyl)propanoic acid (S-allylthio-L-cysteine) were prepared from 3-[(methoxycarbonyl)dithio]-L-alanine, obtained from the reaction of L-cysteine with methoxycarbonylsulfenyl chloride. The occurrence of these S-(+)-alk(en)ylthio-L-cysteine derivatives in onion (Allium cepa L.)
IR, Raman and SERS spectra of 2-(methoxycarbonylmethylsulfanyl)-3, 5-dinitrobenzene carboxylic acid.
Raj A, et al.
Journal of the Brazilian Chemical Society, 20(3), 549-559 (2009)
A SIMPLE AND EFFICIENT SYNTHESIS OF 3-2 PYRIDINYLDITHIO PROPANOIC ACID HYDRAZIDE: A HETEROBIFUNCTIONAL CROSSLINKER.
Saxena R and Manjan MJ.
International Journal of Pharmacy and Pharmaceutical Sciences, 4 (2012)
Mauricio F Erben et al.
Inorganic chemistry, 41(5), 1064-1071 (2002-03-05)
The molecular structure and conformational properties of methoxycarbonylsulfenyl chloride, CH(3)OC(O)SCl, were determinated in the gas and solid phases by gas electron diffraction, low-temperature X-ray diffraction, and vibrational spectroscopy. Furthermore, quantum chemical calculations were performed. Experimental and theoretical methods result in
Mauricio F Erben et al.
The journal of physical chemistry. A, 111(33), 8062-8071 (2007-07-31)
Total ion yield spectra and photoinduced fragmentations following S 2p, Cl 2p, and O 1s inner shell excitations of methoxycarbonylsulfenyl chloride, CH(3)OC(O)SCl, have been studied in the gaseous phase by using synchrotron radiation and multicoincidence techniques, which include photoelectron-photoion coincidence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service