Skip to Content
Merck
All Photos(1)

Key Documents

357782

Sigma-Aldrich

2,3,5-Trichlorobenzoic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€87.30

€87.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€87.30

About This Item

Linear Formula:
Cl3C6H2CO2H
CAS Number:
Molecular Weight:
225.46
Beilstein:
2097064
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€87.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

166-167 °C (lit.)

functional group

carboxylic acid
chloro

SMILES string

OC(=O)c1cc(Cl)cc(Cl)c1Cl

InChI

1S/C7H3Cl3O2/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2H,(H,11,12)

InChI key

CGFDSIZRJWMQPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
D57507345504684155
Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

mp

166-167 °C (lit.)

mp

184-187 °C (lit.)

mp

-

mp

121-125 °C

form

solid

form

powder

form

liquid

form

solid

functional group

carboxylic acid

functional group

-

functional group

acyl chloride, chloro

functional group

carboxylic acid, fluoro

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of temperature on biological degradation of phenols, benzoates and phthalates under methanogenic conditions.
Leven L and Schnurer A.
International Biodeterioration & Biodegradation, 55(2), 153-160 (2005)
Rearrangement of Halotropones. Chloride Exchange in Tribromotropolone.
Doering WE and Knox LH.
Journal of the American Chemical Society, 74(22), 5683-5687 (1952)
Meng Qi et al.
Analytical and bioanalytical chemistry, 412(25), 6995-7006 (2020-08-02)
Dicamba herbicide is increasingly used in the world, in particular' with the widespread cultivation of genetically modified dicamba-resistant crops. However, the drift problem in the field has caused phytotoxicity against naive, sensitive crops, raising legal concerns. Thus, it is particularly
Prakash Karegoudar et al.
European journal of medicinal chemistry, 43(4), 808-815 (2007-09-07)
The reaction of 2,3,5-trichlorobenzoic acid hydrazide with carbon disulfide and potassium hydroxide followed by treatment with hydrazine hydrate afforded 3-(2,3,5-trichlorophenyl)-4-amino-1,2,4-triazole-5-thione (6). Alternatively, this triazole was also synthesized by fusing 2,3,5-trichlorobenzoic acid with thiocarbohydrazide. Condensation of (6) with various aromatic carboxylic
Emeline L Maillet et al.
Journal of medicinal chemistry, 52(21), 6931-6935 (2009-10-13)
We show that phenoxyauxin herbicides and lipid-lowering fibrates inhibit human but not rodent T1R3. T1R3 as a coreceptor in taste cells responds to sweet compounds and amino acids; in endocrine cells of gut and pancreas T1R3 contributes to glucose sensing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service