Skip to Content
Merck
All Photos(1)

Documents

348945

Sigma-Aldrich

cis,cis-Mucononitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH=CHCH=CHCN
CAS Number:
Molecular Weight:
104.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

128-131 °C (lit.)

SMILES string

[H]\C(C#N)=C([H])\C([H])=C(\[H])C#N

InChI

1S/C6H4N2/c7-5-3-1-2-4-6-8/h1-4H/b3-1-,4-2-

InChI key

QLQYLCRMTTXKHS-CCAGOZQPSA-N

General description

cis,cis-Mucononitrile (Z,Z-2,4-hexadienedinitrile) can be prepared by the copper-catalyzed oxidation of o-phenylenediamine. 13C MAS NMR spectra of cis,cis-mucononitrile oriented in liquid-crystalline media has been studied. Pt-catalyzed addition of diethylphosphine to cis,cis-mucononitrile yields the new diphosphine, Et2PCH(CN)CH(CH2CH2CN)PEt2.

Application

cis,cis-Mucononitrile may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regiochemistry of platinum-catalyzed hydrophosphination of a diene. Formation of the chiral diphosphine Et2PCH (CN) CH (CH2CH2CN) PEt2 via monophosphine intermediates.
Kovacik I, et al.
Organometallics, 25(2), 536-539 (2006)
13C MAS NMR Spectra of cis, cis-Mucononitrile in Liquid-Crystalline Media and in the Solid State. Orientation of the Carbon Chemical-Shift Tensors.
Ramanathan KV, et al.
Journal of Magnetic Resonance, 123(2), 196-200 (1996)
Copper-catalyzed oxidation of o-phenylenediamines to cis, cis-mucononitriles.
Kajimoto T, et al.
The Journal of Organic Chemistry, 41(8), 1389-1393 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service