Skip to Content
Merck
All Photos(1)

Documents

347949

Sigma-Aldrich

3-Cyanoindole

98%

Synonym(s):

3-Indolecarbonitrile, NSC 24935

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2
CAS Number:
Molecular Weight:
142.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

179-182 °C (lit.)

SMILES string

N#Cc1c[nH]c2ccccc12

InChI

1S/C9H6N2/c10-5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H

InChI key

CHIFTAQVXHNVRW-UHFFFAOYSA-N

General description

3-Cyanoindole undergoes regiospecific bromination to afford 6-bromo-3-cyanoindole. 3-Cyanoindole is formed during the flash vacuum thermolysis (FVT) of 3-azidoquinoline.

Application

Reactant for preparation of:
  • Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Inhibitors of the C-terminal domain of RNA polymerase II with antitumor activities
  • 4-substituted β-lactams
  • Biologically active Indoles
  • Inhibitors of glycogen synthase kinase 3β (GSK-3)
  • HIV-1 integrase inhibitors
  • Indole fragments as inosine monophosphate dehydrogenase (IMPDH) inhibitors
  • Aziridinomitosene skeleton
  • Potential antiviral agents

Reactant for:
  • Intramolecular oxidative C-H coupling reactions with applications in medium-ring synthesis techniques

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Y Miyake et al.
Organic letters, 2(14), 2121-2123 (2000-07-13)
[reaction: see text] A concise synthesis of topsentin A (R(1) = R(2) = H) and nortopsentins B (R(1) = Br, R(2) = H) and D (R(1) = R(2) = H) is described from oxotryptamine 5 via reduction of acyl cyanide
Interconversion of Nitrenes, Carbenes, and Nitrile Ylides by Ring Expansion, Ring Opening, Ring Contraction, and Ring Closure: 3-Quinolylnitrene, 2-Quinoxalylcarbene, and 3-Quinolylcarbene.
Kvaskoff D, et al.
Australian Journal of Chemistry, 62(3), 275-286 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service