Skip to Content
Merck
All Photos(1)

Key Documents

347175

Sigma-Aldrich

Diethyl phthalimidomalonate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO6
CAS Number:
Molecular Weight:
305.28
Beilstein:
303845
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

mp

73-76 °C (lit.)

functional group

ester
imide

SMILES string

CCOC(=O)C(N1C(=O)c2ccccc2C1=O)C(=O)OCC

InChI

1S/C15H15NO6/c1-3-21-14(19)11(15(20)22-4-2)16-12(17)9-7-5-6-8-10(9)13(16)18/h5-8,11H,3-4H2,1-2H3

InChI key

SZNGBHWKVWEBKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl phthalimidomalonate is an N-malonic ester substituted pthalimide. UV spectra of diethyl phthalimidomalonate has been determined from pH 1 to 12.8.

Application

Diethyl phthalimidomalonate (diethyl 2-phthalimidomalonate) may be employed as suitable neutral carrier in the new poly(vinyl chloride) membrane sensor, for the determination of Cr3+ ions in sample solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence for nc. far.. pi.* transition in N-malonic esters of heterocyclic compounds.
Nnadi JC and Wang SY.
Journal of the American Chemical Society, 92(14), 4421-4425 (1970)
Potentiometric detection of Cr 3+ ions in solution by a chromium (III) electrochemical sensor based on diethyl 2-phthalimidomalonate doped in polymeric membrane.
Zamani HA and Sahebnasagh S.
International Journal of Electrochemical Science, 8, 3708-3720 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service