Skip to Content
Merck
All Photos(1)

Key Documents

347051

Sigma-Aldrich

3-Fluorophenethylamine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
139.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.509 (lit.)

bp

87 °C/15 mmHg (lit.)

density

1.066 g/mL at 25 °C (lit.)

functional group

amine
fluoro

SMILES string

NCCc1cccc(F)c1

InChI

1S/C8H10FN/c9-8-3-1-2-7(6-8)4-5-10/h1-3,6H,4-5,10H2

InChI key

AUCVZEYHEFAWHO-UHFFFAOYSA-N

Application

3-Fluorophenethylamine may be used in the synthesis of:
  • N-(3-florophenyl)ethylcaffeamide and its anti-inflammatory activity was evaluated[1]
  • N-{2-[(3-fluorophenyl)ethyl]}-2-methylpropanamide[2]
  • N-(3′-fluorophenyl)ethyl-4-azahexacyclo[5.4.1.02,6.03,10.05,9.08,11]dodecan-3-ol[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jung-Chun Liao et al.
International journal of molecular sciences, 14(8), 15199-15211 (2013-07-28)
In this study, we evaluated the anti-inflammatory activity of one synthetic product, N-(3-Florophenyl)ethylcaffeamide (abbrev. FECA), by using animal model of λ-carrageenan-induced paw edema in mice. The anti-inflammatory mechanism of FECA was determined by measuring the levels of cyclooxygenase-2 (COX-2), nitric
Susumu Watanuki et al.
Chemical & pharmaceutical bulletin, 59(8), 1029-1037 (2011-08-02)
A series of 1-isopropyl-1,2,3,4-tetrahydroisoquinoline derivatives were synthesized and their bradycardic activities were evaluated in isolated guinea pig right atria. Structure-activity relationship studies revealed that the introduction of an appropriate substituent and its position on the 1,2,3,4-tetrahydroisoquinoline ring are essential for
Xiang Liu et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 1(1), 31-38 (2006-06-23)
Three new trishomocubane analogues based on the 4-azahexacyclo[5.4.1.0(2,6).0(3,10).0(5,9).0(8,11)] dodecane-3-ol skeleton have been synthesised and assessed for their affinities at both sigma-1 and sigma-2 receptors. The effect of various N-substitution on the polycyclic moiety was examined. All synthesised compounds displayed high

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service