Skip to Content
Merck
All Photos(2)

Key Documents

342858

Sigma-Aldrich

Silicon dioxide

fused (pieces), 4 mm, 99.99% trace metals basis

Synonym(s):

Cristobalite, Quartz, Sand, Silica

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

fused (pieces)

refractive index

n20/D 1.544 (lit.)

particle size

4 mm

mp

1610 °C (lit.)

density

2.6 g/mL at 25 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for vacuum deposition.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soania Mathur et al.
Journal of Parkinson's disease, 5(2), 263-268 (2015-02-28)
Despite an urgent need for new medications, clinical trials in Parkinson's have a relatively low rate of success. Although many reasons have been proposed for this, the opinions of patients and scientists, the two principal stakeholders, have not been widely
Yijun Yao et al.
Journal of hazardous materials, 298, 221-231 (2015-06-10)
In 2002, U.S. EPA proposed a general buffer zone of approximately 100 feet (30 m) laterally to determine which buildings to include in vapor intrusion (VI) investigations. However, this screening distance can be threatened by factors such as extensive surface
Keren B Turton et al.
PloS one, 10(5), e0127243-e0127243 (2015-05-20)
Signal transducer and activator of transcription 3 (STAT3) is a key mediator of leukocyte differentiation and proliferation. The 3' end of STAT3 transcripts is subject to two alternative splicing events. One results in either full-length STAT3α or in STAT3β, which
Manuela Piazzi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(4), 1383-1394 (2015-01-01)
The phosphoinositide-dependent signal transduction pathway has been implicated in the control of a variety of biologic processes, such as the regulation of cellular metabolism and homeostasis, cell proliferation and differentiation, and apoptosis. One of the key players in the regulation
Danielle D Cloutier et al.
Applied and environmental microbiology, 81(15), 4904-4913 (2015-05-17)
Microbial communities within beach sand play a key role in nutrient cycling and are important to the nearshore ecosystem function. Escherichia coli and enterococci, two common indicators of fecal pollution, have been shown to persist in the beach sand, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service