Skip to Content
Merck
All Photos(3)

Key Documents

342254

Sigma-Aldrich

Rhenium(IV) oxide

99.7% trace metals basis

Synonym(s):

Rhenium dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€594.00
5 G
€2,150.00

€594.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€594.00
5 G
€2,150.00

About This Item

Linear Formula:
ReO2
CAS Number:
Molecular Weight:
218.21
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

€594.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

99.7% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: rhenium

SMILES string

O=[Re]=O

InChI

1S/2O.Re

InChI key

WXBOMIKEWRRKBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rhenium(IV) oxide is a crystalline solid that adopts a rutile structure. It can be used as a catalyst, and additive in the fabrication of superconducting ceramics. The layers containing ReO2 can be used to protect dielectric layers, inter-connects, and ferroelectric capacitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service