Skip to Content
Merck
All Photos(1)

Key Documents

341231

Sigma-Aldrich

5-(Dimethylaminomethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO4
CAS Number:
Molecular Weight:
199.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

120-123 °C (lit.)

SMILES string

CN(C)\C=C1/C(=O)OC(C)(C)OC1=O

InChI

1S/C9H13NO4/c1-9(2)13-7(11)6(5-10(3)4)8(12)14-9/h5H,1-4H3

InChI key

YKMXFLMJEHHKMU-UHFFFAOYSA-N

General description

5-(Dimethylaminomethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione is a β-dimethyl amino acrylate and its synthesis has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New reagents from ?N, N-Dimethyl amino methoxy methylenium methyl sulphate?-Synthesis of 3-Amino-4-cyano pyrazole.
Arava VR, et al.
Pharmaceutics null
New reagents from ?N, N-Dimethyl amino methoxy methylenium methyl sulphate?-Synthesis of 3-Amino-4-cyano pyrazole.
Arava VR, et al.
Der Pharma Chemica, 2(3), 231-231 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service