Skip to Content
Merck
All Photos(1)

Key Documents

340669

Sigma-Aldrich

8-Fluoro-1-benzosuberone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11FO
CAS Number:
Molecular Weight:
178.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.541 (lit.)

bp

84-86 °C/0.001 mmHg (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc2CCCCC(=O)c2c1

InChI

1S/C11H11FO/c12-9-6-5-8-3-1-2-4-11(13)10(8)7-9/h5-7H,1-4H2

InChI key

INDIENDTRXERIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

8-Fluoro-1-benzosuberone was employed in the synthesis of 9-bromo, 2-fluoro substituted, and Z-ring locked (by a trimethylene bridge at C-8 and C-11) crystalline fluorine-substituted derivative of cinnamaldehyde.[1] It was also used in the synthesis of 9-bromo, 2-fluoro substituted, and Z-ring locked (by a trimethylene bridge at C8 and C11) crystalline fluorine substituted bicyclic ring-fused polyene bearing a δ-bromo-α,β,γ,δ-unsaturated ester unit.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 9-Bromo-2-fluoro-6, 7-dihydro-5H-benzocycloheptene-8-carboxaldehyde.
Silveira MH, et al.
Synthetic Communications, 39(8), 1367-1377 (2009)
Synthesis and Structure of 9-Bromo-2-fluoro-8-[ethyl,(E)-2-propenoate-3-yl]-6, 7-dihydro-5H-benzocycloheptene.
Silveira MH, et al.
Journal of Chemical Crystallography, 41(4), 458-463 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service