Skip to Content
Merck
All Photos(1)

Key Documents

340200

Sigma-Aldrich

3-Amino-5-methylpyrazole

97%

Synonym(s):

5-Methyl-3-pyrazolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N3
CAS Number:
Molecular Weight:
97.12
Beilstein:
1904
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

213 °C/14 mmHg (lit.)

mp

45-47 °C (lit.)

SMILES string

Cc1cc(N)n[nH]1

InChI

1S/C4H7N3/c1-3-2-4(5)7-6-3/h2H,1H3,(H3,5,6,7)

InChI key

FYTLHYRDGXRYEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Complexation of the amino- and carboxyl-protected tripeptide with 3-amino-5-methylpyrazole was studied by low-temperature NMR experiments in a freonic solvent.[1]

Application

3-Amino-5-methylpyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides.[2] It was also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

P Saweczko et al.
Inorganic chemistry, 40(17), 4409-4419 (2001-08-07)
The use of 3-aminopyrazole derivatives as beta-sheet templates is investigated using a series of ferrocenoyl (Fc)-dipeptides (Fc-Gly(2)-OEt, Fc-Ala(2)-OBzl, Fc-Leu-Phe-OMe, Fc-Val-Phe-OMe, Fc-Phe(2)-OMe, Fc-Leu(2)-OMe, Fc-Val(2)-OMe). The synthesis and full characterization are reported. The solid-state structures of Fc-Gly(2)-OMe and Fc-Leu-Phe-OMe show extensive hydrogen
Synthesis of partially hydrogenated pyrazolo [3, 4-b] quinolinones by condensation of 3-amino-5-methylpyrazole with aromatic aldehydes and dimedone.
Lipson VV, et al.
Russ. J. Org. Chem., 42(7), 1015-1021 (2006)
Wei Wang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(3), 854-861 (2006-11-07)
Complexation of the amino- and carboxyl-protected tripeptide Piv-L-Val-L-Val-L-Val-tBu with 3-methylpyrazole and 3-amino-5-methylpyrazole was studied by low-temperature NMR experiments in a freonic solvent. The peptide forms an extended beta-type structure at all temperatures and associates through hydrogen bonding with the two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service