Skip to Content
Merck
All Photos(1)

Key Documents

338222

Sigma-Aldrich

Styrene-d8

≥98 atom % D, ≥98% (CP), contains 4-t-butylcatechol as stabilizer

Synonym(s):

Deuterated styrene, Phenylethylene-D8, Vinylbenzene-D8, Octadeuterostyrene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€175.00
5 G
€551.00

€175.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
€175.00
5 G
€551.00

About This Item

Linear Formula:
C6D5CD=CD2
CAS Number:
Molecular Weight:
112.20
Beilstein:
2327045
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.12

€175.00


Please contact Customer Service for Availability

Request more information

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

liquid

contains

4-t-butylcatechol as stabilizer

refractive index

n20/D 1.5445 (lit.)

bp

145-146 °C (lit.)

mp

-31 °C

density

0.979 g/mL at 25 °C (lit.)

mass shift

M+8

storage temp.

2-8°C

SMILES string

[2H]\C([2H])=C(/[2H])c1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2/i1D2,2D,3D,4D,5D,6D,7D

InChI key

PPBRXRYQALVLMV-GDALLCCDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Styrene-d8 is used as an internal standard for the quantitative analysis of volatile organic hazardous in fuel oil using headspace GC-MS.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Chem. Phys. , 98, 8994-8994 (1993)
Xu-Liang Cao et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(10), 2045-2051 (2018-08-28)
Low levels of styrene may be found in foods as a result of possible migration from polystyrene-based food packaging and as a result of its formation during the biodegradation of a wide variety of naturally occurring compounds with structures similar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service