Skip to Content
Merck
All Photos(1)

Key Documents

337722

Sigma-Aldrich

1,3,5-Tribenzylhexahydro-1,3,5-triazine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27N3
CAS Number:
Molecular Weight:
357.49
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

100 °C/0.005 mmHg (lit.)

mp

49-51 °C (lit.)

functional group

phenyl

SMILES string

C1N(CN(CN1Cc2ccccc2)Cc3ccccc3)Cc4ccccc4

InChI

1S/C24H27N3/c1-4-10-22(11-5-1)16-25-19-26(17-23-12-6-2-7-13-23)21-27(20-25)18-24-14-8-3-9-15-24/h1-15H,16-21H2

InChI key

VWVZIRPJPFJGFE-UHFFFAOYSA-N

Application

1,3,5-Tribenzylhexahydro-1,3,5-triazine was used:
  • in the synthesis of alkyl β-aminocarboxylates via trifluoromethanesulfonic acid catalyzed reaction with ketene silyl acetals[1]
  • as ligand to study the assembling process of a heterogeneous Cr-based single-site ethylene trimerization catalyst by X-ray absorption spectroscopy[2]
  • as ligand during conversion of Phillips Cr/SiO2 polymerisation catalyst to ethylene trimerisation catalyst after assembling new active sites on the silica surface[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina N Nenu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(18), 4756-4763 (2006-04-07)
X-ray absorption spectroscopy at the Cr K- and L(2,3)-edges was used to study the assembling process of a heterogeneous Cr-based single-site catalyst. The starting point was a Phillips-type system with monochromate species anchored on a silica surface, which was first
Turning a Cr-based heterogeneous ethylene polymerisation catalyst into a selective ethylene trimerisation catalyst.
Nenu CN, et al.
J. Mol. Catal. A: Chem., 269(1), 5-11 (2007)
Trifluoromethanesulfonic acid-promoted reaction of hexahydro-1, 3, 5-triazines with ketene silyl acetals. convenient synthesis of alkyl ?-aminocarboxylates.
Ikeda K, et al.
Tetrahedron Letters, 24(9), 913-916 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service