Skip to Content
Merck
All Photos(2)

Key Documents

33737

Sigma-Aldrich

Dibenzyl N,N-dimethylphosphoramidite

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€245.00

€245.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€245.00

About This Item

Empirical Formula (Hill Notation):
C16H20NO2P
CAS Number:
Molecular Weight:
289.31
Beilstein:
7427974
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€245.00


Please contact Customer Service for Availability

Request a Bulk Order

refractive index

n20/D 1.553

density

1.089 g/mL at 20 °C (lit.)

functional group

amine
phenyl

storage temp.

2-8°C

SMILES string

CN(C)P(OCc1ccccc1)OCc2ccccc2

InChI

1S/C16H20NO2P/c1-17(2)20(18-13-15-9-5-3-6-10-15)19-14-16-11-7-4-8-12-16/h3-12H,13-14H2,1-2H3

InChI key

GDEJGFHRPXLQNN-UHFFFAOYSA-N

Application

Dibenzyl N,N-dimethylphosphoramidite was used in the preparation of dibenzyl phosphoramidates incorporating the representative C-terminal protected amino acids leucine, phenylalanine, glutamic acid and proline.[1] It was also used for the fast and clean phosphitylation of alcohols, including hindered alcohols.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of N-phosphoryl amino acids via phosphoramidite amine-exchange.
Chow CP and Berkman CE.
Tetrahedron Letters, 39(41), 7471-7474 (1998)
M. Schneider et al.
Ger. Offen. 4'329'533 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service