Skip to Content
Merck
All Photos(2)

Key Documents

336718

Sigma-Aldrich

Cadmium bromide tetrahydrate

98%

Synonym(s):

Cadmium dibromide tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdBr2 · 4H2O
CAS Number:
Molecular Weight:
344.28
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
solid

Assay

98%

form

solid

reaction suitability

reagent type: catalyst
core: cadmium

SMILES string

O.O.O.O.Br[Cd]Br

InChI

1S/2BrH.Cd.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

PANYFCXNZSEXSU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Cadmium bromide tetrahydrate can be used in the synthesis of cadmium zinc sulfate (CdZnS) based on quantum dots. It can also be used in the synthesis of coordination polymers, which can be used as second-order non-linear optical (NLO) materials.[1][2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Crystal structure of catena-poly [[cadmium (II)-di-\mu2-bromido-\mu2-l-proline-\kappa2O: O?] monohydrate]
Sathiskumar S, et al.
Acta Crystallographica Section E, Structure Reports Online, 71(2), 217-219 (2015)
Aqueous synthesis of Cu-doped CdZnS quantum dots with controlled and efficient photoluminescence
Yakoubi A, et al.
Journal of Luminescence, 175(2), 193-202 (2016)
Marion Dufour et al.
ACS nano, 13(5), 5326-5334 (2019-04-13)
Zinc blende II-VI semiconductor nanoplatelets (NPLs) are defined at the atomic scale along the thickness of the nanoparticle and are initially capped with carboxylates on the top and bottom [001] facets. These ligands are exchanged on CdSe NPLs with halides

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service