Skip to Content
Merck
All Photos(2)

Key Documents

336556

Sigma-Aldrich

Bis(triphenylsilyl) chromate

96%

Synonym(s):

Bis(triphenylsilyl) ester chromic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3SiO]2CrO2
CAS Number:
Molecular Weight:
634.79
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 336556, please contact your local Merck office or dealer. Contact Technical Service

Assay

96%

reaction suitability

reagent type: oxidant

mp

159 °C (dec.) (lit.)

SMILES string

O=[Cr](=O)(O[Si](c1ccccc1)(c2ccccc2)c3ccccc3)O[Si](c4ccccc4)(c5ccccc5)c6ccccc6

InChI

1S/2C18H15OSi.Cr.2O/c2*19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;;;/q2*-1;+2;;

InChI key

AQLZCGLPNYEIDH-UHFFFAOYSA-N

Application

Bis(triphenylsilyl) chromate [(Ph3SiO)2] can be used as a catalyst:
  • For ethylene polymerization.[1]
  • For the conversion of secondary benzylic and allylic trimethylsilyl ethers to ketones by oxidation reaction using tert-butyl hydroperoxide.[2]

It can also be used to prepare supported catalysts with silica gel or montmorillonite K-10. These catalysts can be used in the oxidation of alcohols to corresponding carbonyl compounds.[3][4]
Reactant for the XPS and XRD characterization of chromium-silica catalysts

Silyl chromate catalyst for
  • Olefin polymerization and ethylene polymerization
  • Benzylic oxidations used as a phase transfer catalyst

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethylene polymerization with supported bis (triphenylsilyl) chromate catalysts
Carrick WL, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 10(9), 2609-2620 (1972)
Oxidation of Alcohols by Silica Gel-Supported Bis (Trimethylsilyl) Chromate Under Microwave Irradiation Without Solvent
Heravi MM, et al.
Synthetic Communications, 29(2), 163-166 (1999)
Bis (triphenylsilyl) chromate catalyzed oxidation of silyl ethers by tert-butyl hydroperoxide
Muzart J and Ajjou A
Synlett, 1991(07), 497-498 (1991)
Oxidation of alcohols by montmorillonite K-10 supported bis (trimethylsilyl) chromate
Heravi MM, et al.
Monatshefte fur Chemie / Chemical Monthly, 129(12), 1305-1308 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service