Skip to Content
Merck
All Photos(1)

Key Documents

33562

Sigma-Aldrich

N,N′-Dibenzoyl-L-cystine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€337.00

€337.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€337.00

About This Item

Empirical Formula (Hill Notation):
C20H20N2O6S2
CAS Number:
Molecular Weight:
448.51
Beilstein:
2068210
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€337.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

powder

optical activity

[α]20/D −215±8°, c = 1% in ethanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

195-200 °C (dec.)

application(s)

peptide synthesis

SMILES string

OC(=O)[C@H](CSSC[C@H](NC(=O)c1ccccc1)C(O)=O)NC(=O)c2ccccc2

InChI

1S/C20H20N2O6S2/c23-17(13-7-3-1-4-8-13)21-15(19(25)26)11-29-30-12-16(20(27)28)22-18(24)14-9-5-2-6-10-14/h1-10,15-16H,11-12H2,(H,21,23)(H,22,24)(H,25,26)(H,27,28)/t15-,16-/m0/s1

InChI key

GUTXMPQWQSOAIY-HOTGVXAUSA-N

Application

Reactant involved in:
  • Synthesis of integrin antagonists
  • Dissipative self-assembly of molecular gelator using chemical fuel
  • Synthesis of crosslinkable hydrogel for proliferation of encapsulated human derm fibroblast
  • Analysis for use as metallo-β-lactamase inhibitors
  • Amplification of bifunctional disulfide ligands for calmodulin

Other Notes

Chiral modifier of LiBH4 for enantioselective reductions of aromatic, heteroaromatic and unsaturated ketones to alcohols[1][2][3][4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Chemical Communications, 801-801 (1987)
Journal of the Chemical Society. Chemical Communications, 138-138 (1985)
K. Soai et al.
Journal of the Chemical Society. Chemical Communications, 413-413 (1984)
P.G. Sammes et al.
Journal of the Chemical Society. Perkin Transactions 1, 111-111 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service