Skip to Content
Merck
All Photos(1)

Documents

332925

Sigma-Aldrich

4-Chlorobenzenesulfonic acid

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SO3H
CAS Number:
Molecular Weight:
192.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

impurities

<3.5% H2SO4

bp

149 °C/22 mmHg (lit.)

SMILES string

OS(=O)(=O)c1ccc(Cl)cc1

InChI

1S/C6H5ClO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,(H,8,9,10)

InChI key

RJWBTWIBUIGANW-UHFFFAOYSA-N

General description

4-Chlorobenzenesulfonic acid is the major polar by-product formed during the chemical synthesis of 1,1,1-trichloro-2,2-bis-(4-chlorophenyl)ethane (DDT).

Application

4-Chlorobenzenesulfonic acid was employed as anion dopants for the matrix-assisted laser desorption/ionization-mass spectrometry (MALDI-MS) of neutral oligosaccharides.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A W Wong et al.
Analytical chemistry, 72(7), 1419-1425 (2000-04-14)
Alkylsulfonates are examined as anion dopants for the matrix-assisted laser desorption/ionization-mass spectrometry (MALDI-MS) of neutral oligosaccharides. The anion dopants allow neutral oligosaccharides to be examined in the same mixture as acidic oligosaccharides. The alkylsulfonate dopants interact strongly with the oligosaccharide
Rafael Blasco et al.
Environmental microbiology, 10(6), 1591-1600 (2008-03-12)
Pseudomonas aeruginosa RW41 is the first bacterial strain, which could be isolated by virtue of its capability to mineralize 4-chlorobenzenesulfonic acid (4CBSA), the major polar by-product of the chemical synthesis of 1,1,1-trichloro-2,2-bis-(4-chlorophenyl)ethane (DDT). This capability makes the isolate a promising

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service