Skip to Content
Merck
All Photos(1)

Key Documents

332798

Sigma-Aldrich

2,3,4,5-Tetrafluoronitrobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC6F4NO2
CAS Number:
Molecular Weight:
195.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.473 (lit.)

bp

80 °C/24 mmHg (lit.)

density

1.618 g/mL at 25 °C (lit.)

functional group

fluoro
nitro

SMILES string

[O-][N+](=O)c1cc(F)c(F)c(F)c1F

InChI

1S/C6HF4NO2/c7-2-1-3(11(12)13)5(9)6(10)4(2)8/h1H

InChI key

MKMDVNZEIQDZEP-UHFFFAOYSA-N

General description

2,3,4,5-Tetrafluoronitrobenzene is a polyfluoroarene.[1] Mercuration of 2,3,4,5-tetrafluoronitrobenzene has been reported.[2] It reacts with thioureas to afford para substituted diaryl sulphides.[1] The nucleophilic aromatic substitution reactions of 2,3,4,5-tetrafluoronitrobenzene with methanol has been reported.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aromatic polyfluoro-compounds-LIII reactions of polyflouroarenes with thioureas.
Coe PL, et al.
Tetrahedron, 28(1), 105-109 (1972)
Organomercury compounds: XVII. The mercuration of 1, 2, 3, 4-tetrafluorobenzene, 1, 2, 3, 5-tetrafluorobenzene, 2, 3, 5, 6-tetrafluoroanisole, and 2, 3, 4, 5-tetrafluoronitrobenzene.
Albrecht HB and Deacon GB.
Journal of Organometallic Chemistry, 57(1), 77-86 (1973)
Reductively activated ?Polar? nucleophilic aromatic substitution. III: The reactions of polyfluoronitrobenzenes with methanol.
Niat M, et al.
Tetrahedron Letters, 35(48), 9059-9062 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service