Skip to Content
Merck
All Photos(1)

Key Documents

332062

Sigma-Aldrich

Dichlorobis(tributylphosphine)nickel(II)

Synonym(s):

NSC 54233

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
{[CH3(CH2)3]3P}2NiCl2
CAS Number:
Molecular Weight:
534.23
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

reaction suitability

core: nickel
reagent type: catalyst

mp

43-45 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl[Ni]Cl.CCCCP(CCCC)CCCC.CCCCP(CCCC)CCCC

InChI

1S/2C12H27P.2ClH.Ni/c2*1-4-7-10-13(11-8-5-2)12-9-6-3;;;/h2*4-12H2,1-3H3;2*1H;/q;;;;+2/p-2

InChI key

KPFOFXWYYUNJOZ-UHFFFAOYSA-L

Application

Catalyst for:
  • Wenkert arylations
  • Regioselective [2+2+2] cycloadditions
  • Polymerization reactions

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service