Skip to Content
Merck
All Photos(3)

Key Documents

331449

Sigma-Aldrich

3-Amino-5-hydroxypyrazole

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€112.00

€112.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€112.00

About This Item

Empirical Formula (Hill Notation):
C3H5N3O
CAS Number:
Molecular Weight:
99.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€112.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

mp

214 °C (dec.) (lit.)

SMILES string

Nc1cc(O)[nH]n1

InChI

1S/C3H5N3O/c4-2-1-3(7)6-5-2/h1H,(H4,4,5,6,7)

InChI key

QZBGOTVBHYKUDS-UHFFFAOYSA-N

General description

Molecular and vibrational structure of 3-amino-5-hydroxypyrazole was investigated by density functional method.[1]

Application

3-Amino-5-hydroxypyrazole was used in the synthesis of 2,6-difluoro-N-(3-methoxy-1H-pyrazolo[3,4-b]pyridine-5-yl)-3-(propylsulfonamidio)benzamide[2] and heteroaromatic 3-hydroxy-5,6-diphenylpyrazolo[3,4-b]pyridines.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Swaminathan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(5), 1776-1782 (2008-09-02)
In this work, we report a combined experimental and theoretical study on molecular and vibrational structure of 3-amino-5-hydroxypyrazole (3A5HP). The Fourier transform infrared and Fourier transform Raman spectra of 3A5HP were recorded in the solid phase. The molecular geometry and
Zun-Ting Zhang et al.
Journal of combinatorial chemistry, 12(4), 600-603 (2010-06-08)
A new concise, facile, and efficient method for the synthesis of 3-hydroxy-5,6-diphenylpyrazolo[3,4-b]pyridine derivatives has been described. The cyclocondensation of isoflavones with 3-amino-5-hydroxypyrazole in the presence of sodium methoxide gave fused heteroaromatic 3-hydroxy-5,6-diphenylpyrazolo[3,4-b]pyridines in moderate to good yields.
Min Wang et al.
Bioorganic & medicinal chemistry letters, 23(4), 1017-1021 (2013-01-09)
The authentic standard 2,6-difluoro-N-(3-methoxy-1H-pyrazolo[3,4-b]pyridine-5-yl)-3-(propylsulfonamidio)benzamide was synthesized from 2,6-difluorobenzoic acid and 3-amino-5-hydroxypyrazole in 9 steps with 1% overall chemical yield. Direct desmethylation of the reference standard with TMSCl/NaI gave the precursor 2,6-difluoro-N-(3-hydroxy-1H-pyrazolo[3,4-b]pyridine-5-yl)-3-(propylsulfonamidio)benzamide for radiolabeling in 70% yield. The target tracer 2,6-difluoro-N-(3-[(11)C]methoxy-1H-pyrazolo[3,4-b]pyridine-5-yl)-3-(propylsulfonamidio)benzamide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service