Skip to Content
Merck
All Photos(1)

Key Documents

331228

Sigma-Aldrich

Anisole-(methyl-d3)

99 atom % D

Synonym(s):

Methoxy-d3-benzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€131.00

€131.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
€131.00

About This Item

Linear Formula:
C6H5OCD3
CAS Number:
Molecular Weight:
111.16
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€131.00


Please contact Customer Service for Availability

Request more information

vapor pressure

5.25 mmHg ( 50 °C)

Quality Level

isotopic purity

99 atom % D

form

liquid

refractive index

n20/D 1.5164 (lit.)

bp

154 °C (lit.)

mp

-37 °C (lit.)

density

1.023 g/mL at 25 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])Oc1ccccc1

InChI

1S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6H,1H3/i1D3

InChI key

RDOXTESZEPMUJZ-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service