Skip to Content
Merck
All Photos(1)

Documents

330671

Sigma-Aldrich

2-(Trimethylsilyl)ethoxymethyl-triphenylphosphonium chloride

98%

Synonym(s):

SEM-triphenylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2CH2OCH2P(C6H5)3Cl
CAS Number:
Molecular Weight:
429.01
Beilstein:
4632507
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

145-149 °C (lit.)

SMILES string

[Cl-].C[Si](C)(C)CCOC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C24H30OPSi.ClH/c1-27(2,3)20-19-25-21-26(22-13-7-4-8-14-22,23-15-9-5-10-16-23)24-17-11-6-12-18-24;/h4-18H,19-21H2,1-3H3;1H/q+1;/p-1

InChI key

NEUMNYXEDIPGJD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Enantioselective synthesis of (-)-sarain A using asymmetric Michael addition, cyclization, ring-closing metathesis, intramolecular Stille coupling, and late-state installation of tertiary amine-aldehyde proximity interaction
  • Preparation of aliphatic and alicyclic aldehydes by homologation of aldehydes
  • Wittig reactions
  • Preparation of optically active (1S,5R)-(+)-3-azabicyclo[3.2.2]non-6-en-2-one via enantioselective intramolecular cyclopropanation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service