Skip to Content
Merck
All Photos(1)

Key Documents

330604

Sigma-Aldrich

2-Methyl-1-cyclohexanecarboxylic acid, mixture of cis and trans

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H10CO2H
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.4633 (lit.)

bp

241-242 °C/746 mmHg (lit.)

density

1.009 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CC1CCCCC1C(O)=O

InChI

1S/C8H14O2/c1-6-4-2-3-5-7(6)8(9)10/h6-7H,2-5H2,1H3,(H,9,10)

InChI key

VSKXJRZPVDLHFY-UHFFFAOYSA-N

Application

2-Methyl-1-cyclohexanecarboxylic acid was used in the synthesis of substituted cyclohexyl carbonyl chlorides.[1]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and evaluation of mefway analogs as ligands for serotonin 5HT1A receptors.
Thio JP, et al.
Medicinal Chemistry Research, 1-7 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service