Skip to Content
Merck
All Photos(1)

Key Documents

329606

Sigma-Aldrich

Isosafrol

mixture of cis and trans

Synonym(s):

1,2-Methylenedioxy-4-propenylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
Beilstein:
82640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.573 (lit.)

bp

77-86 °C/3.5 mmHg (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

C\C=C\c1ccc2OCOc2c1

InChI

1S/C10H10O2/c1-2-3-8-4-5-9-10(6-8)12-7-11-9/h2-6H,7H2,1H3/b3-2+

InChI key

VHVOLFRBFDOUSH-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isosafrol is also referred as 1,2-methylenedioxy-4(1-propenyl)benzene. It is an important intermediate for the synthesis of drugs like L-DOPA.[1] Peracid oxidation of isosafrole yields isomers of 2,4-dimethyl-3,5-bis(3,4-methylenedioxyphenyl)tetrahydrofuran.[2] Encapsulated vanadyl compounds in Y-zeolite pores catalyzed isosafrol oxidation under microwave irradiation was reported.[3]

Application

Isosafrol was used in preparation of 1-(3,4-methylenedioxyphenyl)-2-propanone (MDP-2-P or PMK).[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P H Jellinck et al.
The Journal of steroid biochemistry and molecular biology, 46(6), 791-798 (1993-12-01)
The effect of indole-3-carbinol (IC), an anticarcinogen present in cruciferous vegetables, to alter the metabolism of 4-androstenedione (AD) by female rat liver microsomes was investigated and compared to that of its main gastric conversion product, diindolylmethane (DIM) as well as
M J Leaver et al.
Molecular marine biology and biotechnology, 2(6), 338-345 (1993-12-01)
A full length cDNA coding for cytochrome P450 1A1 was isolated from a plaice liver cDNA library constructed in lambda ZAPII. The deduced amino acid sequence of this cDNA was 78% homologous to that of rainbow trout P450 1A1 and
C L Miranda et al.
Toxicology and applied pharmacology, 142(1), 123-132 (1997-01-01)
Liver microsomes from juvenile trout metabolized DMBA to unknown highly polar metabolites (X) and to DMBA-t-5,6-diol, DMBA-t-8,9-diol, 7-OHM-12-MBA, 7M-12-OHMBA, 2-OH-DMBA, 4-OH-DMBA, and trace amounts of DMBA-t-3,4-diol. Treatment of trout with beta-naphthoflavone (BNF) and isosafrole (ISF) increased the formation of these
D Ronisz et al.
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology, 121(1-3), 289-296 (1999-02-11)
The CYP1A enzyme in liver of rainbow trout (Oncorhynchus mykiss) and eelpout (Zoarces viviparus) was induced by intraperitoneal injections of isosafrole (ISF), beta-naphthoflavone (BNF), retene, 3,3',4,4'-tetrachlorobiphenyl (TCB), Clophen A50 and combinations of these compounds. The livers were sampled 5 days
D Taras-Valéro et al.
Environmental and molecular mutagenesis, 32(4), 314-324 (1999-01-09)
Mice of the XVIInc/Z and DBA/2N strains, which are responsive and nonresponsive, respectively, to the aryl hydrocarbon (Ah) receptor, were treated with the hepatocarcinogen 5,9-dimethyldibenzo[c,g]carbazole and their livers were examined by nuclease P1-enhanced 32P-postlabeling for the levels of DNA adducts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service