Skip to Content
Merck
All Photos(2)

Key Documents

329495

Sigma-Aldrich

1,4,7-Triazacyclononane trihydrochloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€142.00

€142.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€142.00

About This Item

Empirical Formula (Hill Notation):
C6H15N3 · 3HCl
CAS Number:
Molecular Weight:
238.59
Beilstein:
5287631
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€142.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

288 °C (dec.) (lit.)

SMILES string

Cl[H].Cl[H].Cl[H].C1CNCCNCCN1

InChI

1S/C6H15N3.3ClH/c1-2-8-5-6-9-4-3-7-1;;;/h7-9H,1-6H2;3*1H

InChI key

HNPMVNQYFPWBKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4,7-Triazacyclononane trihydrochloride can be used as a reagent to prepare:
  • Nonadentate ligand, 1,4,7-tris[(6-carboxypyridin-2-yl)methyl]-1,4,7-triazacyclononane (H3tpatcn), which is used to prepare lanthanide complexes with high water solubility and rigid C3 symmetric structures.[1]
  • Dansyl cryptands as fluorescent indicators via amination of (bromobenzyl)triazacycloalkane and oxadiamines.[2]
  • [1,4,7-tri(3-butenyl)-1,4,7-triazacyclononane and 1,4,7-tri(2-propenyl)-1,4,7-triazacyclononane] by reacting with corresponding alkenyl halide.[3]

Possible reagent for compleximetric titrations with high cation-binding selectivity.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Dansyl-Substituted Cryptands Containing Triazacycloalkane Moieties and their Evaluation as Fluorescent Chemosensors
Chernichenko NM, et al.
Synlett, 28(20), 2800-2806 (2017)
An investigation into alkenyl-functionalized 1, 4, 7-triazacyclononanes: Synthesis, metal complexation, and attempted olefin metathesis
Baker MV, et al.
Australian Journal of Chemistry, 55(10), 655-660 (2002)
Solid-state and solution properties of the lanthanide complexes of a new nonadentate tripodal ligand derived from 1, 4, 7-triazacyclononane
Gateau C, et al.
Dalton Transactions, 2428-2433 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service