Skip to Content
Merck
All Photos(1)

Key Documents

329320

Sigma-Aldrich

Bromoform-d

≥99.5 atom % D, ≥99% (CP), contains copper as stabilizer

Synonym(s):

Tribromomethane-d

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€147.00
25 G
€252.00

€147.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
10 G
€147.00
25 G
€252.00

About This Item

Linear Formula:
CDBr3
CAS Number:
Molecular Weight:
253.74
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

€147.00


Please contact Customer Service for Availability

Request more information

isotopic purity

≥99.5 atom % D

Quality Level

Assay

≥99% (CP)

form

liquid

contains

copper as stabilizer

impurities

~1% carbon tetrabromide

refractive index

n20/D 1.596 (lit.)

bp

136.5 °C (lit.)

density

2.906 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]C(Br)(Br)Br

InChI

1S/CHBr3/c2-1(3)4/h1H/i1D

InChI key

DIKBFYAXUHHXCS-MICDWDOJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao He et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(12), 1295-1309 (2020-04-16)
The crossed molecular beam reactions of the methylidyne radical (CH; X2 Π) with 1,3-butadiene (CH2 CHCHCH2 ; X1 Ag ) along with their (partially) deuterated counterparts were performed at collision energies of 20.8 kJ mol-1 under single collision conditions. Combining our laboratory
Chao He et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(48), 30142-30150 (2020-11-18)
Since the postulation of carbenes by Buchner (1903) and Staudinger (1912) as electron-deficient transient species carrying a divalent carbon atom, carbenes have emerged as key reactive intermediates in organic synthesis and in molecular mass growth processes leading eventually to carbonaceous

Questions

  1. Hello, I would like to know the concentration of copper in bromoform-d, the Sigma-Aldrich product n° 329320. Best regards

    1 answer
    1. Copper is added to this product as a stabilizer and is present at a range of 1-2%.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service