Skip to Content
Merck
All Photos(1)

Key Documents

321273

Sigma-Aldrich

Ethane

10 ppm in nitrogen

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH3
CAS Number:
Molecular Weight:
30.07
Beilstein:
1730716
MDL number:

vapor density

1.05 (vs air)

vapor pressure

37.95 atm ( 21.1 °C)

autoignition temp.

881 °F

expl. lim.

13 %

concentration

10 ppm in nitrogen

bp

−88 °C (lit.)

mp

−172 °C (lit.)

density

0.362 g/mL at 20 °C (lit.)

SMILES string

CC

InChI

1S/C2H6/c1-2/h1-2H3

InChI key

OTMSDBZUPAUEDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For calibrating analytical instruments.

Packaging

Packaged in lightweight no-return cylinder

Recommended products

Control valve Z166995 or regulator Z167002 is recommended.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Aguado et al.
Journal of the American Chemical Society, 134(36), 14635-14637 (2012-08-24)
Absolute ethylene/ethane separation is achieved by ethane exclusion on silver-exchanged zeolite A adsorbent. This molecular sieving type separation is attributed to the pore size of the adsorbent, which falls between ethylene and ethane kinetic diameters.
Isobel J Simpson et al.
Nature, 488(7412), 490-494 (2012-08-24)
After methane, ethane is the most abundant hydrocarbon in the remote atmosphere. It is a precursor to tropospheric ozone and it influences the atmosphere's oxidative capacity through its reaction with the hydroxyl radical, ethane's primary atmospheric sink. Here we present
Reika Kanya et al.
The Journal of chemical physics, 136(20), 204309-204309 (2012-06-07)
Two-body Coulomb explosion processes of ethane (CH(3)CH(3)) and its isotopomers (CD(3)CD(3) and CH(3)CD(3)) induced by an intense laser field (800 nm, 1.0 × 10(14) W/cm(2)) with three different pulse durations (40 fs, 80 fs, and 120 fs) are investigated by
Paul O Wennberg et al.
Environmental science & technology, 46(17), 9282-9289 (2012-08-03)
We use historical and new atmospheric trace gas observations to refine the estimated source of methane (CH(4)) emitted into California's South Coast Air Basin (the larger Los Angeles metropolitan region). Referenced to the California Air Resources Board (CARB) CO emissions
Ramiro F Quijano-Quiñones et al.
Molecules (Basel, Switzerland), 17(4), 4661-4671 (2012-04-24)
The energy change on each Occupied Molecular Orbital as a function of rotation about the C-C bond in ethane was studied using the B3LYP, mPWB95 functional and MP2 methods with different basis sets. Also, the effect of the ZPE on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service