320595
Butadiene sulfone-2,2,5,5-d4
98 atom % D
Synonym(s):
2,2,5,5-Tetradeutero-3-sulfolene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
isotopic purity
98 atom % D
form
solid
mp
67-68 °C (lit.)
mass shift
M+4
SMILES string
[2H]C1([2H])C=CC([2H])([2H])S1(=O)=O
InChI
1S/C4H6O2S/c5-7(6)3-1-2-4-7/h1-2H,3-4H2/i3D2,4D2
InChI key
MBDNRNMVTZADMQ-KHORGVISSA-N
Application
Convenient source of butadiene-1,1,4,4-d4.
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
233.6 °F - closed cup
Flash Point(C)
112.00 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of the Chemical Society. Chemical Communications, 1005-1005 (1987)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service